di-Lithiumtetraborat ≥99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Lithiumtetraborat
13419.18 13419.30 13419.A1
13419.18EA 41.1 EUR
13419.18 13419.30 13419.A1
di-Lithiumtetraborat ≥99% (Metall-Basis)
di-Lithiumtetraborat
Purity: 99% CAS No.: 12007-60-2 Molecular Formula: B4Li2O7 Formula Weight: 169.12 Physical Form: Powder Density: 2.4 Appearance: White Odour: Odourless Melting Point: 930° Merck Reference: 14,5524 MDL No.: MFCD00011083
Formel: Li₂B₄O₇
Molecular Weight: 169.12 g/mol
Schmelzpunkt: 930 °C
Dichte: 2.4
Lagertemperatur: Raumtemperatur
MDL: MFCD00011083
CAS-Nummer: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,5524