Lithiumtetrafluoroborat

Lieferant: FLUOROCHEM

002848-25G 002848-100G
FLUO002848-25GEA 82.1 EUR
FLUO002848-25G FLUO002848-100G
Lithiumtetrafluoroborat
Lithium tetrafluoroborate
Formel: BF₄Li
Molecular Weight: 93.74 g/mol
Dichte: 0.852 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011087
CAS-Nummer: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR