Natriumtetrafluoroborat ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Sodium fluoroborate
11562.36
11562.A3
11562.36EA
49.8
EUR
11562.36
11562.A3
Natriumtetrafluoroborat ≥95%
Sodium tetrafluoroborate
Purity: 95%
CAS No.: 13755-29-8
Molecular Formula: BF4Na
Formula Weight: 109.79
Physical Form: Lump/Granular
Density: 2.47
Appearance: White to off-white
Melting Point: 384° dec.
Solubility: Soluble in water
Merck Reference: 14,8617
MDL No.: MFCD00003515
Fluorinating agent
Formel:
BF₄Na Molecular Weight: 109,79 g/mol Schmelzpunkt: 384 °C (Zersetzung) Dichte: 2,47 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003515 CAS-Nummer: 13755-29-8 EINECS: 237-340-6 Merck Index: 14,8617 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...