Natriumtetrafluoroborat
Lieferant: Honeywell Chemicals
Synonyme:
Sodium fluoroborate
01508.9050
01508.9025
HONE01508.9050EA
0
EUR
HONE01508.9050
HONE01508.9025
Natriumtetrafluoroborat
Sodium tetrafluoroborate
Formel:
BF₄Na Molecular Weight: 109.79 g/mol Schmelzpunkt: 384 Dichte: ∼2.47 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003515 CAS-Nummer: 13755-29-8 EINECS: 237-340-6 Merck Index: 12,08761 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...