Lithiummetaborat ≥99,997% (Metall-Basis), Puratronic®
Lieferant: Thermo Fisher Scientific
Achtung
10739.36 10739.14 10739.22
10739.36EA 1074 EUR
10739.36 10739.14 10739.22
Lithiummetaborat ≥99,997% (Metall-Basis), Puratronic®
Lithium metaborate
Purity: 99.997% CAS No.: 13453-69-5 Molecular Formula: BLiO2 Formula Weight: 49.75 Physical Form: Powder Density: 1.397 Appearance: White Odour: Odourless Melting Point: 845° MDL No.: MFCD00011089
Formel: BLiO₂
Molecular Weight: 49.75 g/mol
Siedepunkt: 1719
Schmelzpunkt: 845
Dichte: 1.397
Lagertemperatur: Raumtemperatur
MDL: MFCD00011089
CAS-Nummer: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546