Lithiummetaborat, wasserfrei ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
Achtung
12591.18 12591.30 12591.A1
12591.18EA 50.3 EUR
12591.18 12591.30 12591.A1
Lithiummetaborat, wasserfrei ≥99,9% (Metall-Basis)
Lithium metaborate
Purity: 99.9% CAS No.: 13453-69-5 Molecular Formula: BLiO2 Formula Weight: 49.75 Physical Form: Powder Density: 1.397 Appearance: White Odour: Odourless Melting Point: 845° MDL No.: MFCD00011089
Formel: BLiO₂
Molecular Weight: 49.75 g/mol
Siedepunkt: 1719
Schmelzpunkt: 845
Dichte: 1.397
Lagertemperatur: Raumtemperatur
MDL: MFCD00011089
CAS-Nummer: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546