Kaliumbromid ≥99% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bromkalium
12612.18
12612.30
12612.A1
12612.A7
12612.18EA
27.6
EUR
12612.18
12612.30
12612.A1
12612.A7
Kaliumbromid ≥99% ACS
Kaliumbromid
Purity: 99% min
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Physical Form: Crystalline
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 730°
Boiling Point: 1435°
Solubility: Freely soluble in water. Soluble in alcohol, glycerol
Merck Reference: 14,7618
MDL No.: MFCD00011358
Manufacturing of photographic papers, process engraving and lithography, spectroscopy, as an analytical lab reagent
Formel:
KBr Molecular Weight: 119 g/mol Siedepunkt: 1435 °C Schmelzpunkt: 730 Dichte: 2.75 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011358 CAS-Nummer: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |