Lithiumbromid, wasserfrei ≥99%
Lieferant: Thermo Scientific
Achtung
13408.22 13408.36
13408.22EA 48.2 EUR
13408.22 13408.36
Lithiumbromid, wasserfrei ≥99%
Lithium bromide
Purity: 99% min CAS No.: 7550-35-8 Molecular Formula: BrLi Formula Weight: 86.85 Physical Form: Crystalline Density: 3.464 Appearance: White Odour: Odourless Melting Point: 550° Boiling Point: 1265° Solubility: Very soluble in water and alcohol. Soluble in methanol, acetone, and glycol. Slightly soluble in pyridine Merck Reference: 14,5526 MDL No.: MFCD00011077
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
Formel: LiBr
Molecular Weight: 86,84 g/mol
Siedepunkt: 1265 °C
Schmelzpunkt: 550 °C
Dichte: 3,464
Lagertemperatur: Raumtemperatur
MDL: MFCD00011077
CAS-Nummer: 7550-35-8
EINECS: 231-439-8
Merck Index: 14,5526