Natriumbromid ≥99,9955% (Metall-Basis), Puratronic®
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
Bromnatrium
10860.14
10860.22
10860.14EA
142
EUR
10860.14
10860.22
Natriumbromid ≥99,9955% (Metall-Basis), Puratronic®
Natriumbromid
Purity: 99.9955%
CAS No.: 7647-15-6
Molecular Formula: NaBr
Formula Weight: 102.89
Physical Form: Powder
Density: 3.203
Refractive Index: 1.6412
Sensitivity: Hygroscopic
Melting Point: 755°
Boiling Point: 1390°
Merck Reference: 14,8594
MDL No.: MFCD00003475
Formel:
NaBr Molecular Weight: 102,89 g/mol Siedepunkt: 1390 °C Schmelzpunkt: 755 °C Dichte: 3,203 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003475 CAS-Nummer: 7647-15-6 EINECS: 231-599-9 Merck Index: 14,8594 |