Natriumbromid ≥99,99% (Metall-Basis)
Lieferant: Thermo Scientific
Achtung
Synonyme: Bromnatrium
87604.06 87604.14
87604.06EA 51 EUR
87604.06 87604.14
Natriumbromid ≥99,99% (Metall-Basis)
Natriumbromid
Purity: 99.99% CAS No.: 7647-15-6 Molecular Formula: NaBr Formula Weight: 102.89 Physical Form: Crystalline Density: 3.203 Refractive Index: 1.6412 Sensitivity: Hygroscopic Melting Point: 755° Boiling Point: 1390° Merck Reference: 14,8594 MDL No.: MFCD00003475
Formel: NaBr
Molecular Weight: 102,89 g/mol
Siedepunkt: 1390 °C
Schmelzpunkt: 755 °C
Dichte: 3,203
Lagertemperatur: Raumtemperatur
MDL: MFCD00003475
CAS-Nummer: 7647-15-6
EINECS: 231-599-9
Merck Index: 14,8594