Kaliumbromat 99+%, reinst
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
208855000
208850025
ACRO208855000EA
70.8
EUR
ACRO208855000
ACRO208850025
Kaliumbromat 99+%, reinst
Potassium bromate
Formel:
KBrO₃ Molecular Weight: 167 g/mol Schmelzpunkt: 350 °C Dichte: 3.27 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011359 CAS-Nummer: 7758-01-2 EINECS: 231-829-8 UN: 1484 ADR: 5.1 II Merck Index: 12,07779 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder and/or chunks |
Titration Iodometric | ≥99.0 % |
Bromide (Br) | ≤500 ppm |
Sulfate (SO4) | ≤100 ppm |
Lead (Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...