Kaliumhydrogencarbonat ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: Kaliumbicarbonat

A12429.0C A12429.0E A12429.36
A12429.0CEA 299 EUR
A12429.0C A12429.0E A12429.36
Kaliumhydrogencarbonat ≥99%
Kaliumhydrogencarbonat

Purity: 99% CAS No.: 298-14-6 Molecular Formula: KHCO3 Formula Weight: 100.12 Odour: Odourless Merck Reference: 14,7609 MDL No.: MFCD00011402

Formel: KHCO₃
Molecular Weight: 100,12 g/mol
Schmelzpunkt: 292 °C
Dichte: 2,17
Lagertemperatur: Raumtemperatur
MDL: MFCD00011402
CAS-Nummer: 298-14-6
EINECS: 206-059-0
Merck Index: 14,7609

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR