Cäsiumchlorid, dry_ultra 99,9 Metals basis
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Cesium Chloride
ampuled under argon
42832.06 42832.14 42832.22
42832.06EA 87.5 EUR
42832.06 42832.14 42832.22
Cäsiumchlorid, dry_ultra 99,9 Metals basis
Cesium chloride
Purity: 99.9% CAS No.: 7647-17-8 Molecular Formula: CsCl Formula Weight: 168.36 Physical Form: Powder, Ampouled under argon Density: 3.99 Refractive Index: 1.6418 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 646° Boiling Point: 1303° Merck Reference: 14,2011 MDL No.: MFCD00010955
Formel: CsCl
Molecular Weight: 168.36 g/mol
Siedepunkt: 1303 °C
Schmelzpunkt: 646
Dichte: 3.99
Lagertemperatur: Raumtemperatur
MDL: MFCD00010955
CAS-Nummer: 7647-17-8
EINECS: 231-600-2
Merck Index: 14,2011