Rubidiumchlorid ≥99,8% (Metall-Basis)

Lieferant: Thermo Scientific
Achtung

Synonyme: Rubidiumchloride

10563.09 10563.18
10563.09EA 110 EUR
10563.09 10563.18
Rubidiumchlorid ≥99,8% (Metall-Basis)
Rubidium chloride

Purity: 99.8% CAS No.: 7791-11-9 Molecular Formula: ClRb Formula Weight: 120.92 Physical Form: -20 Mesh Crystalline Density: 2.800 Refractive Index: 1.493 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 718° Boiling Point: 1390° Merck Reference: 14,8284 MDL No.: MFCD00011187

Formel: RbCl
Molecular Weight: 120,92 g/mol
Siedepunkt: 1390 °C
Schmelzpunkt: 718 °C
Dichte: 2,800
Lagertemperatur: Raumtemperatur
MDL: MFCD00011187
CAS-Nummer: 7791-11-9
EINECS: 232-240-9
Merck Index: 14,8284

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR