Kaliumfluorid, wasserfrei 99%

Lieferant: Thermo Fisher Scientific
Gefahr

14130.18 14130.30 14130.A1
14130.18EA 38.7 EUR
14130.18 14130.30 14130.A1
Kaliumfluorid, wasserfrei 99%
Potassium fluoride

Purity: 99% CAS No.: 7789-23-3 Molecular Formula: FK Formula Weight: 58.10 Physical Form: Crystalline Density: 2.48 Refractive Index: 1.363 Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 859° Boiling Point: 1505° Beilstein Reference: 3902818 Merck Reference: 14,7632 UN No.: 1812 MDL No.: MFCD00011398

Formel: KF
Siedepunkt: 1505 °C
Schmelzpunkt: 859 °C
Dichte: 2,48
Lagertemperatur: Raumtemperatur
MDL: MFCD00011398
CAS-Nummer: 7789-23-3
EINECS: 232-151-5
UN: 1812
ADR: 6.1,III
Merck Index: 14,7632

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR