Kaliumfluorid ≥99% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42216.22
42216.36
42216.22EA
39.2
EUR
42216.22
42216.36
Kaliumfluorid ≥99% ACS
Potassium fluoride
Purity: 99% min
CAS No.: 7789-23-3
Molecular Formula: FK
Formula Weight: 58.10
Physical Form: Powder
Density: 2.48
Refractive Index: 1.363
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 859°
Boiling Point: 1505°
Solubility: Solubility in water at 21° (96g/100ml). Freely soluble in boiling water. Soluble in aqueous HF, liquid NH3. Insoluble in anhydrous alcohol
Beilstein Reference: 3902818
Merck Reference: 14,7632
UN No.: 1812
MDL No.: MFCD00011398
Formel:
KF Siedepunkt: 1505 °C Schmelzpunkt: 859 °C Dichte: 2,48 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011398 CAS-Nummer: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 14,7632 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...