Lithiumfluorid ≥99,99% (Metall-Basis), Puratronic®

Lieferant: Thermo Scientific
Gefahr

10736.03 10736.09 10736.18 10736.30
10736.03EA 39.3 EUR
10736.03 10736.09 10736.18 10736.30
Lithiumfluorid ≥99,99% (Metall-Basis), Puratronic®
Lithium fluoride

Purity: 99.99% CAS No.: 7789-24-4 Molecular Formula: FLi Formula Weight: 25.94 Physical Form: Powder Density: 2.635 Refractive Index: 1.3915 Appearance: White Odour: Odourless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090

Formel: FLi
Molecular Weight: 25,94 g/mol
Siedepunkt: 1680 °C
Schmelzpunkt: 845 °C
Dichte: 2,635
Lagertemperatur: Raumtemperatur
MDL: MFCD00011090
CAS-Nummer: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5531

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR