Lithiumfluorid ≥99,98% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
10986.14 10986.22
10986.14EA 143 EUR
10986.14 10986.22
Lithiumfluorid ≥99,98% (Metall-Basis)
Lithium fluoride
Purity: 99.98% CAS No.: 7789-24-4 Molecular Formula: FLi Formula Weight: 25.94 Physical Form: Powder Density: 2.635 Refractive Index: 1.3915 Appearance: White Odour: Odourless Melting Point: 845° Boiling Point: 1680° Merck Reference: 14,5531 UN No.: 3288 MDL No.: MFCD00011090
Formel: FLi
Molecular Weight: 25,94 g/mol
Siedepunkt: 1680 °C
Schmelzpunkt: 845 °C
Dichte: 2,635
Lagertemperatur: Raumtemperatur
MDL: MFCD00011090
CAS-Nummer: 7789-24-4
EINECS: 232-152-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5531