Lithiumhydrid ≥99,4% (Metall-Basis)

Lieferant: Thermo Scientific
Gefahr

41596.14 41596.22
41596.14EA 114 EUR
41596.14 41596.22
Lithiumhydrid ≥99,4% (Metall-Basis)
Lithium hydride

Purity: 99.4% CAS No.: 7580-67-8 Molecular Formula: HLi Formula Weight: 7.95 Physical Form: Powder Density: 0.78 Appearance: White to grey Odour: Odourless Melting Point: 686° Merck Reference: 14,5533 UN No.: 1414 MDL No.: MFCD00011074

Formel: HLi
Molecular Weight: 7,95 g/mol
Schmelzpunkt: 686 °C
Dichte: 0,78
Lagertemperatur: Raumtemperatur
MDL: MFCD00011074
CAS-Nummer: 7580-67-8
EINECS: 231-484-3
UN: 1414
ADR: 4.3,I
Merck Index: 14,5533

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR