Natriumiodid ≥99,5% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Iodnatrium
40196.22
40196.36
40196.22EA
64.6
EUR
40196.22
40196.36
Natriumiodid ≥99,5% ACS
Natriumiodid
Purity: 99.5% min
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Physical Form: Crystalline Powder
Density: 3.67
Refractive Index: 1.7745
Sensitivity: Hygroscopic
Odour: Odourless
Melting Point: 651°
Boiling Point: 1300-1304°
Solubility: Soluble in water, alcohol, glycerol, acetone. Insoluble in ether
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formel:
NaI Molecular Weight: 149.89 g/mol Siedepunkt: 1300…1304 °C Schmelzpunkt: 651 °C Dichte: 3.67 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003532 CAS-Nummer: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9 III Merck Index: 14,8631 |