Natriumiodat 99%, Pulver, reinst Reagens

Lieferant: Thermo Fisher Scientific
Gefahr

419560050 419565000
ACRO419560050EA 24.7 EUR
ACRO419560050 ACRO419565000
Natriumiodat 99%, Pulver, reinst Reagens
Natriumiodat
Formel: INaO₃
Molecular Weight: 197.89 g/mol
Schmelzpunkt: 560 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003533
CAS-Nummer: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1 II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Titration Iodometric ≥98.5 %
pH 5.0 to 8.0 (5 % at 20°C)
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR