Kaliumcyanat ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Cyansäure Kaliumsalz
33356.22
33356.36
33356.A3
33356.22EA
34.6
EUR
33356.22
33356.36
33356.A3
Kaliumcyanat ≥97%
Kaliumcyanat
Purity: 97%
CAS No.: 590-28-3
Molecular Formula: KOCN
Formula Weight: 81.12
Physical Form: Crystalline
Density: 2.056
Appearance: White
Odour: Odourless
Melting Point: 700-900° dec.
Solubility: Soluble in water. Slightly soluble in alcohol
Merck Reference: 14,7625
MDL No.: MFCD00011411
Formel:
CKNO Molecular Weight: 81,12 g/mol Schmelzpunkt: 700…900 °C (Zersetzung) Dichte: 2,056 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011411 CAS-Nummer: 590-28-3 EINECS: 209-676-3 Merck Index: 14,7625 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...