Natriumhexafluorophosphat
Lieferant: FLUOROCHEM
002866-10G
002866-25G
002866-100G
FLUO002866-10GEA
60.9
EUR
FLUO002866-10G
FLUO002866-25G
FLUO002866-100G
Natriumhexafluorophosphat
Sodium hexafluorophosphate
Formel:
F₆NaP Molecular Weight: 167,95 g/mol Schmelzpunkt: 450 °C Dichte: 2,505 g/cm³ (20 °C) |
MDL:
MFCD00011122 CAS-Nummer: 21324-39-0 EINECS: 244-333-1 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...