Dinatriumtetrachloropalladat Hydrat (30% Pd) ≥99,95% (Metall-Basis)
Lieferant: Thermo Scientific
Gefahr
Synonyme: Natriumtetrachloropalladat(II), Natrium-Palladium(II)-chlorid
11886.04 11886.09 11886.18
11886.04EA 277 EUR
11886.04 11886.09 11886.18
Dinatriumtetrachloropalladat Hydrat (30% Pd) ≥99,95% (Metall-Basis)
Dinatriumtetrachloropalladat Hydrat
Purity: 99.95% CAS No.: 13820-53-6 Molecular Formula: Cl4Na2Pd.xH2O (x≈3) Formula Weight: 294.20(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red-brown MDL No.: MFCD00003487
Formel: Cl₄Na₂Pd·xH₂O
Molecular Weight: 294,21 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003487
CAS-Nummer: 13820-53-6
EINECS: 237-502-6