Natriumsulfit 95,0-100,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Lieferant: VWR Chemicals
28130.260EA
38.2
EUR
28130.260
28130.292
28130.460
Natriumsulfit 95,0-100,5%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Sodium sulphite
Formel:
Na₂SO₃ Molecular Weight: 126,04 g/mol Schmelzpunkt: > 500 °C Dichte: 2,63 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003503 CAS-Nummer: 7757-83-7 EINECS: 231-821-4 REACH: 01-2119537420-49 Merck Index: 13,08757 |
Spezifikation Testergebnisse
Assay (at filling) | 95.0 - 100.5 % |
Appearance of solution | Passes test Ph.Eur. |
Identification A | Passes test Ph.Eur. |
Identification B | Passes test Ph.Eur. |
Identification C | Passes test Ph.Eur. |
Solution S1 | Passes test Ph.Eur. |
Colouration (10 %; water) | Max. 10 APHA |
Heavy metals (as Pb) | Max. 10 ppm |
S2O3 (Thiosulphate) | Max. 0.1 % |
Fe (Iron) | Max. 10 ppm |
Se (Selenium) | Max. 10 ppm |
Zn (Zinc) | Max. 25 ppm |
Conforms to Reag. Ph.Eur. | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...