Sodium tellurate(VI) hydrate (≥45% Te)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
75106.09
75106.18
75106.09EA
75.4
EUR
75106.09
75106.18
Sodium tellurate(VI) hydrate (≥45% Te)
di-Natriumtellurtetraoxid Hydrat
Sodium tellurite is an intermediate in the extraction of the element, tellurium; it is a product obtained from anode slimes and is a precursor to tellurium. Solutions of sodium tellurite are used for black or blue-black coatings on iron, steel, aluminum, and copper.
CAS No.: 10101-83-4
Molecular Formula: Na2TeO4.xH2O
Formula Weight: 237.58(anhy)
Physical Form: Crystalline
Melting Point: dec.
Merck Reference: 14,8686
MDL No.: MFCD00014242
Formel:
Na₂O₄Te·xH₂O Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014242 CAS-Nummer: 10101-83-4 EINECS: 233-259-5 UN: 3284 ADR: 6.1,II Merck Index: 14,8686 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...