Natriumhexafluorotitanat

Lieferant: FLUOROCHEM

007283-25G 007283-100G
FLUO007283-25GEA 0 EUR
FLUO007283-25G FLUO007283-100G
Natriumhexafluorotitanat
Natriumhexafluorotitanat
Formel: F₆Na₂Ti
MDL: MFCD00014244
CAS-Nummer: 17116-13-1
EINECS: 241-181-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR