Strontiumchlorid, wasserfrei ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
12202.36
12202.A3
12202.36EA
79.8
EUR
12202.36
12202.A3
Strontiumchlorid, wasserfrei ≥95%
Strontium chloride
Purity: 95%
CAS No.: 10476-85-4
Molecular Formula: SrCl2
Formula Weight: 158.53
Physical Form: Powder
Density: 3.052
Refractive Index: 1.650
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 873°
Boiling Point: 1250°
Merck Reference: 14,8840
MDL No.: MFCD00011249
Formel:
Cl₂Sr Molecular Weight: 158,53 g/mol Siedepunkt: 1250 °C Schmelzpunkt: 873 °C Dichte: 3,052 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011249 CAS-Nummer: 10476-85-4 EINECS: 233-971-6 Merck Index: 14,8840 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...