Ethoxymethylenmalononitril 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
117921000
117925000
ACRO117921000EA
97.8
EUR
ACRO117921000
ACRO117925000
Ethoxymethylenmalononitril 98%
Ethoxymethylenmalononitril
Formel:
C₆H₆N₂O Molecular Weight: 122.13 g/mol Siedepunkt: 160 °C (12 mmHg) Schmelzpunkt: 63…69 °C Flammpunkt: 155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001854 CAS-Nummer: 123-06-8 EINECS: 204-597-0 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Yellow to orange-brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 63 to 68 °C |
GC | ≥97.5 % |
Manganese (Mn) | ≤2 ppm |
Heavy metals (as Pb) | ≤0.001 % |
Insoluble matter | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...