2-Mercapto-2-thiazolin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Thiazoline-2-Thiol, 4,5-Dihydrothiazole-2-thiol
125560050
125561000
125565000
ACRO125560050EA
22.8
EUR
ACRO125560050
ACRO125561000
ACRO125565000
2-Mercapto-2-thiazolin 98%
2-Mercapto-2-thiazolin
Formel:
C₃H₅NS₂ Molecular Weight: 119.21 g/mol Schmelzpunkt: 103…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126013 CAS-Nummer: 96-53-7 EINECS: 202-512-1 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 106 °C |
Titration with TBAH | 97.5 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...