2-Thioxo-4-imidazolidinon 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Thioxoimidazolidin-4-one
138760050
138760250
138761000
ACRO138760050EA
29.1
EUR
ACRO138760050
ACRO138760250
ACRO138761000
2-Thioxo-4-imidazolidinon 98%
2-Thiohydantoin
Formel:
C₃H₄N₂OS Molecular Weight: 116,14 g/mol Schmelzpunkt: 229…231 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005277 CAS-Nummer: 503-87-7 EINECS: 207-977-4 |
Spezifikation Testergebnisse
Appearance | Cream to yellow to beige to brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...