Aminoguanidiniumhydrogencarbonat 98.5%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Aminoguanidine hydrogen carbonate, Aminoguanidine bicarbonate, 1-Aminoguanidinium hydrogen carbonate, Aminoguanidine hydrogencarbonate, Guanylhydrazine hydrogencarbonate

149060010 149062500 149060051
ACRO149060010EA 73.1 EUR
ACRO149060010 ACRO149062500 ACRO149060051
Aminoguanidiniumhydrogencarbonat 98.5%
Aminoguanidinium hydrogen carbonate
Formel: C₂H₈N₄O₃
Molecular Weight: 136.11 g/mol
Schmelzpunkt: 125 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012949
CAS-Nummer: 2582-30-1
EINECS: 219-956-7
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.3 %
Titration with HClO4 98.0 to 102.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR