N,N-Dimethylglycin Hydrochlorid 99%

Lieferant: Thermo Scientific

Synonyme: N,N-Dimethylglycine HCl

204660050 204660250 204661000
ACRO204660050EA 34.4 EUR
ACRO204660050 ACRO204660250 ACRO204661000
N,N-Dimethylglycin Hydrochlorid 99%
N,N-Dimethylglycin Hydrochlorid
Formel: C₄H₁₀ClNO₂
Molecular Weight: 139,58 g/mol
Schmelzpunkt: 188…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012610
CAS-Nummer: 2491-06-7
EINECS: 219-648-2

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 188 to 198 °C
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR