Diethyl-3-oxoglutarat 95%

Lieferant: Thermo Scientific

Synonyme: Diethyl 1,3-acetonedicarboxylate, Diethyl acetone-1,3-dicarboxylate

325630500 325632500
ACRO325630500EA 60 EUR
ACRO325630500 ACRO325632500
Diethyl-3-oxoglutarat 95%
Diethyl-3-oxoglutarat
Formel: C₉H₁₄O₅
Molecular Weight: 202,21 g/mol
Flammpunkt: 86 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00009200
CAS-Nummer: 105-50-0
EINECS: 203-302-2

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4420 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR