1-(1-Cyclohexen-1-yl)pyrrolidin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
334830250
334831000
ACRO334830250EA
56.7
EUR
ACRO334830250
ACRO334831000
1-(1-Cyclohexen-1-yl)pyrrolidin 95%
1-(1-Cyclohexen-1-yl)pyrrolidin
Formel:
C₁₀H₁₇N Siedepunkt: 114…115 °C (15 mmHg) Flammpunkt: 66 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00003163 CAS-Nummer: 1125-99-1 EINECS: 214-414-6 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow-brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5200 to 1.5240 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...