(S)-(-)-Hydroxypyrrolidin 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-(-)-3-Pyrrolidinol, (S)-Pyrrolidin-3-ol, (S)-(-)-3-Hydroxypyrrolidine
339430010
339430050
ACRO339430010EA
68.4
EUR
ACRO339430010
ACRO339430050
(S)-(-)-Hydroxypyrrolidin 98+%
(S)-(-)-Hydroxypyrrolidin
Formel:
C₄H₉NO Molecular Weight: 87,12 g/mol Siedepunkt: 108…110 °C (8 mmHg) Dichte: 1,048 g/cm³ Flammpunkt: >110 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00192426 CAS-Nummer: 100243-39-8 |
Spezifikation Testergebnisse
Appearance | Colorless to white to yellow Clear liquid or low melting solid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4870 to 1.4910 (20°C, 589 nm) |
GC | ≥98.0 % |
Specific optical rotation | -5° to -8° (20°C, 589 nm) (c=3.5, CH3OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...