N,N-Dimethylglycin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

345200050 345200250
ACRO345200050EA 80.9 EUR
ACRO345200050 ACRO345200250
N,N-Dimethylglycin 97%
N,N-Dimethylglycin
Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
Schmelzpunkt: 175…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004283
CAS-Nummer: 1118-68-9
EINECS: 214-267-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 175 to 181 °C
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR