Pentaethylenglycol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3,6,9,12-Tetraoxatetradecane-1,14-diol
350730050
350730250
ACRO350730050EA
124
EUR
ACRO350730050
ACRO350730250
Pentaethylenglycol 98%
Pentaethylenglycol
Formel:
C₁₀H₂₂O₆ Molecular Weight: 238,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002878 CAS-Nummer: 4792-15-8 EINECS: 225-341-4 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4610 to 1.4630 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...