Methyl-6-aminonicotinat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 6-aminonicotinate, 6-Aminonicotinic acid methyl ester

366810050 366810250
ACRO366810050EA 122 EUR
ACRO366810050 ACRO366810250
Methyl-6-aminonicotinat 97%
Methyl-6-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 159…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00797844
CAS-Nummer: 36052-24-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light beige to pink Powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR