Aminoguanidin Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

368910250 368911000
ACRO368910250EA 56.1 EUR
ACRO368910250 ACRO368911000
Aminoguanidin Hydrochlorid 98%
Aminoguanidin Hydrochlorid
Formel: CH₇ClN₄
Schmelzpunkt: >160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039074
CAS-Nummer: 1937-19-5
EINECS: 217-707-7

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point ≥160 °C
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR