4-Nitrobenzylchlorformiat 95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Nitrobenzyl carbonochloridate
376040050
376040250
ACRO376040050EA
49.9
EUR
ACRO376040050
ACRO376040250
4-Nitrobenzylchlorformiat 95%
4-Nitrobenzylchlorformiat
Formel:
C₈H₆ClNO₄ Molecular Weight: 215,59 g/mol Schmelzpunkt: 32 °C Flammpunkt: 113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00007375 CAS-Nummer: 4457-32-3 EINECS: 224-708-6 UN: 2923 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | Off-white to yellow to beige Solid and/or chunks or liquid after melting |
Infrared spectrum | Conforms |
Melting point | 30 to 36 °C |
Titration Argentometric | ≥94 % (ex Cl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...