Bis(cyclopentadienyl)ruthenium (≥43,2% Ru)
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ruthenocene, Bis(cyclopentadienyl)ruthenium(II)
unter Argon abgefüllt
12781.03
12781.06
12781.03EA
225
EUR
12781.03
12781.06
Bis(cyclopentadienyl)ruthenium (≥43,2% Ru)
Bis(cyclopentadienyl)ruthenium
CAS No.: 1287-13-4
Molecular Formula: C10H10Ru
Formula Weight: 231.26
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Melting Point: 194-198°
MDL No.: MFCD00001442
Formel:
C₁₀Ru Molecular Weight: 221,18 g/mol Schmelzpunkt: 194…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001442 CAS-Nummer: 1287-13-4 EINECS: 215-065-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...