Zirkonium(IV)acetylacetonat
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Tetra(2,4-pentandionato-O,O')zirkonium(IV)
14065.18
14065.30
14065.18EA
94.5
EUR
14065.18
14065.30
Zirkonium(IV)acetylacetonat
Zirkonium(IV)acetylacetonat
CAS No.: 17501-44-9
Molecular Formula: C20H28O8Zr
Formula Weight: 487.66
Physical Form: Crystals
Appearance: White
Odour: Weak
MDL No.: MFCD00000036
Formel:
C₂₀H₂₈O₈Zr Molecular Weight: 487,66 g/mol Schmelzpunkt: 191…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000036 CAS-Nummer: 17501-44-9 EINECS: 241-510-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...