Natrium-3-nitrobenzolsulfonat, (max. 2% H₂O) 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt

A10244.0E A10244.22 A10244.36
A10244.0EEA 241 EUR
A10244.0E A10244.22 A10244.36
Natrium-3-nitrobenzolsulfonat, (max. 2% H₂O) 98%
Natrium-3-nitrobenzolsulfonat

Purity: 98% CAS No.: 127-68-4 Molecular Formula: C6H4NNaO5S.xH2O Formula Weight: 225.16(anhy) Melting Point: 350° MDL No.: MFCD00007490

Formel: C₆H₄NNaO₅S
Molecular Weight: 225,16 g/mol
Schmelzpunkt: 350 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007490
CAS-Nummer: 127-68-4
EINECS: 204-857-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR