Aminoacetaldehyddiethylacetal 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2,2-Diethoxyethanamine, 2-Aminoacetaldehyde diethylacetal
A10427.14
A10427.22
A10427.14EA
64.8
EUR
A10427.14
A10427.22
Aminoacetaldehyddiethylacetal 98%
Aminoacetaldehyddiethylacetal
Purity: 98%
CAS No.: 645-36-3
Molecular Formula: C6H15NO2
Formula Weight: 133.19
Flash Point: 45°(113°F)
Refractive Index: 1.4170
Boiling Point: 162-163°
Beilstein Reference: 506067
UN No.: 2733
MDL No.: MFCD00008136
Formel:
C₆H₁₅NO₂ Molecular Weight: 133,19 g/mol Siedepunkt: 162…163 °C Dichte: 0,916 Flammpunkt: 45 °C (113 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008136 CAS-Nummer: 645-36-3 EINECS: 211-439-4 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...