(±)-3,3-Dimethyl-2-butylamin 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,2,2-Trimethylpropylamine, 3,3-Dimethylbutan-2-amine
A11530.03
A11530.06
A11530.14
A11530.03EA
44
EUR
A11530.03
A11530.06
A11530.14
(±)-3,3-Dimethyl-2-butylamin 98%
(±)-3,3-Dimethyl-2-butylamin
Purity: 98%
CAS No.: 3850-30-4
Molecular Formula: C6H15N
Formula Weight: 101.19
UN No.: 2733
MDL No.: MFCD00008078
Formel:
C₆H₁₅N Molecular Weight: 101,19 g/mol Dichte: 0,755 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008078 CAS-Nummer: 3850-30-4 EINECS: 223-353-4 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...