N-Methylformanilid ≥99%

Lieferant: Thermo Scientific
Achtung

A11829.0C A11829.0E A11829.30 A11829.36 AB117923.0250
A11829.0CEA 1024 EUR
A11829.0C A11829.0E A11829.30 A11829.36 ABCRAB117923.0250
N-Methylformanilid ≥99%
N-Methylformanilid

Purity: 99% CAS No.: 93-61-8 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 126°(258°F) Refractive Index: 1.5610 Melting Point: 9-13° Boiling Point: 242-244° Beilstein Reference: 636496 MDL No.: MFCD00003283

Formel: C₈H₉NO
Molecular Weight: 135,17 g/mol
Siedepunkt: 242…244 °C
Schmelzpunkt: 9…13 °C
Dichte: 1,095
Flammpunkt: 126 °C (258 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003283
CAS-Nummer: 93-61-8
EINECS: 202-262-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR