Hydantoin ≥99%

Lieferant: Thermo Scientific

Synonyme: Imidazolidine-2,4-dione

A12486.0E A12486.22 A12486.36 AB117064.0100
A12486.0EEA 367 EUR
A12486.0E A12486.22 A12486.36 ABCRAB117064.0100
Hydantoin ≥99%
Hydantoin

Purity: 99% CAS No.: 461-72-3 Molecular Formula: C3H4N2O2 Formula Weight: 100.08 Melting Point: 221-223° Beilstein Reference: 110598 Merck Reference: 14,4761 MDL No.: MFCD00005259

Formel: C₃H₄N₂O₂
Molecular Weight: 100,08 g/mol
Schmelzpunkt: 221…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005259
CAS-Nummer: 461-72-3
EINECS: 207-313-3
Merck Index: 14,4761

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR