1,3-Dichlor-5,5-dimethylhydantoin ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,3-Dichloro-5,5-dimethylhydantoine, 1,3-Dichloro-5,5-dimethylimidazolidine-2,4-dione
A12718.0E
A12718.22
A12718.36
A12718.0EEA
178
EUR
A12718.0E
A12718.22
A12718.36
1,3-Dichlor-5,5-dimethylhydantoin ≥98%
1,3-Dichlor-5,5-dimethylhydantoin
Purity: 98%
CAS No.: 118-52-5
Molecular Formula: C5H6Cl2N2O2
Formula Weight: 197.02
Flash Point: 174°(345°F)
Melting Point: 130-134°
Beilstein Reference: 146013
Merck Reference: 14,3065
UN No.: 1479
MDL No.: MFCD00003190
Formel:
C₅H₆Cl₂N₂O₂ Molecular Weight: 197,02 g/mol Schmelzpunkt: 130…134 °C Dichte: 1,500 Flammpunkt: 174 °C (345 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003190 CAS-Nummer: 118-52-5 EINECS: 204-258-7 UN: 3085 ADR: 5.1,II Merck Index: 14,3065 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...