Benzylidenaceton ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Phenylbut-3-en-2-one, 4-Phenyl-3-buten-2-one
A13537.0B
A13537.30
A13537.0BEA
79.5
EUR
A13537.0B
A13537.30
Benzylidenaceton ≥98%
Benzylidenaceton
Purity: 98+%
CAS No.: 122-57-6
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point: 116°(241°F)
Physical Form: Crystalline
Density: 1.038
Sensitivity: Light Sensitive
Odour: Characteristic
Melting Point: 38-42°
Boiling Point: 261°
Beilstein Reference: 742046
Merck Reference: 14,1137
MDL No.: MFCD00008779
Formel:
C₁₀H₁₀O Molecular Weight: 146,19 g/mol Siedepunkt: 261 °C Schmelzpunkt: 38…42 °C Dichte: 1,038 Flammpunkt: 116 °C (241 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008779 CAS-Nummer: 122-57-6 EINECS: 204-555-1 Merck Index: 14,1137 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...